Chemical Duty Diaphragm Vacuum Pumps
Actual product may vary
Fisherbrand
Chemical Duty Diaphragm Vacuum Pumps
Use these pumps for a wide range of laboratory applications. Fisherbrand™ Chemical Duty Diaphragm Vacuum Pumps are ideal for applications involving aggressive solvents and acidic vapors.
Have Questions?
Catalog NumberMaterial
FKS12991131PTFE
Catalog number FKS12991131
Price (SGD)
9,562.00
Each
Material:
PTFE
Price (SGD)
9,562.00
Each
  • Chemical resistant
  • Ideal for rotary evaporator, vacuum concentrator and vacuum oven use to 2mbar
  • Replacement for rotary vane pumps and water aspirators, especially in rough vacuum applications where corrosive vapors are encountered
  • Pump heads are made from PTFE with carbon fibre reinforcing to handle corrosive and aggressive vapors without damage
  • Pumping chambers are hermetically separated from drive space, ensuring extended life of mechanical parts
  • Pumps use PFTE diaphragms sandwiched to strong non-wetted backing and PEEK valves, which are excellent chemical resistance and are extremely hard wearing when compared to elastomer valves
  • Gas ballast valve helps to preserve pump performance when working with condensable vapors
  • Hose nozzles for 8mm I. D. tubing
  • Vibration damping feet
  • Max overpressure; 1 bar
  • IN/EX hose connector: Hose nozzle DNB
  • Motor power: 90W
  • Type of protection (motor): IP54
Available Accessories: 
  • Fisherbrand Vacuum Regulator with Dial Gauge (700458-10)
  • Fisherbrand VCpro601, Vacuum Control Box, 1100 - 1 mbar (600100)
Specifications
Max. Vacuum2 mbar
Electrical Requirements230 V 50/60 Hz motor with Schuko and UK power leads
Dimensions (L x W x H)20 x 26 x 15 cm (7.8 x 10.2 x 5.9 in.)
MaterialPTFE
Noise Level<44db (A), at 50Hz
Weight (English)22.7 lb.
Weight (Metric)10.3 kg
Pump TypeDiaphragm Vacuum Pump
Air Displacement33 L/min.
No. of Pump Heads4
ApplicationsRotary Evaporation
No. of Stages3
Maximum Pressure< 2 mbar
Unit SizeEach