Check Order Status
Rulers
Fisherbrand
Rulers
Moulded in white plastic.
Have Questions?
Catalog Number
Scale Type
FKS12989074
Metric
Catalog number
FKS12989074
Price (SGD)
210.00
PK
Add to cart
Scale Type:
Metric
Price (SGD)
210.00
PK
Add to cart
6mm thick by 25mm wide
1 meter long
Foil printed in black
Specifications
Product Type
Ruler
Color
White
Length (Metric)
1 m
Scale Type
Metric
Thickness
6 mm
Width (Metric)
25 mm
Material
Plastic
Quantity
10 Pack