3M™ Comply™ Indicator Tape
Actual product may vary
Fisherbrand
3M™ Comply™ Indicator Tape
Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)
FKS1298697655 m x 18 mm
FKS1176135155 m x 24 mm
Catalog number FKS12986976
Price (SGD)
25.00
Each
Dimensions (L x W):
55 m x 18 mm
Price (SGD)
25.00
Each
Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.
As an exposure monitor, 3M™ indicator tapes securely seal packs and enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. The chemical indicator ink, printed in a diagonal striped pattern, changes colour or turns darker when exposed to the sterilisation process.
  • Lead-free steam indicator tape is designed for use on all wraps
  • Reliable achesive adheres to packaging material
  • Sustainable, solvent-free manufacturing proces
  • Does not require hazardous waste disposal
  • Redesigned packaging uses up to 30% less plastic
  • Convenient, individually wrapped, easy-to-open rolls
  • Lead free and no added latex
Specifications
Certifications/ComplianceMeets NF EN 867-1 Class A, ISO 11140-1 Class 1
ColorBeige
Dimensions (L x W)55 m x 18 mm
For Use With (Application)Sterilization verification
For Use With (Equipment)Autoclave
Length (Metric)55 m
Product TypeIndicator Tape
FeaturesLead-free, aggressive, pressure sensitive adhesive
Quantity1 Ea.
Sterilization IndicatorYes
Sterilization MethodSteam
Width (Metric) Exterior18 mm
Unit SizeEach