Omnifit™ Connectors with Valve
Actual product may vary
Fisherbrand
Omnifit™ Connectors with Valve
2-way, 3-way Hex, 3-way single key (T), and large-to-small universal connectors with valves
Have Questions?
Catalog NumberType
FKS129750613-way Connector w/Single Key Valve
Catalog number FKS12975061
Price (SGD)
1,098.00
Each
Type:
3-way Connector w/Single Key Valve
Price (SGD)
1,098.00
Each
  • Instant tube connection
  • Chemically insert all PTFE body
  • 50 psi pressure rating
  • On-off manual valve operation
2- and 3-way Connectors with Valves
  • Include Viton™ O-rings, 1/16 in. (1.5mm) PTFE cones, polypropylene caps with internal thread that accepts 1/4 in.-28 UNF fittings
  • PTFE cone provides a flat bottom for the 1/4 in.-28 UNF fitting to give a leak-free connection
  • 1.5mm bore size
  • Accept tubing from 0.5 to 4mm O.D.
  • OF001101 has an inline shut-off valve
  • OF001102 has individual shut-off valves for each of the three lines
  • OF001109 has a diverting valve to allow connection of one line to either of the other two
Large-to-Small Universal Connector with Valve
  • Large end has a Viton seal; small end is supplied with one large and two small Viton O-rings
  • Cap at small end has internal thread that accepts 1/4 in.-28 UNF fittings
  • Accepts 1 to 11mm O.D. tubing
  • 1.5mm bore end
Specifications
ActuationManual
ModeSingle Mode
Bore Size (English)0.05 in.
Closure TypeOmnifit Cap
Bore Size (Metric)0.15 cm
IncludesValve Single key (T)
MaterialPTFE
Operating Pressure50 psi
Port Style0.25 in. -28
Type3-way Connector w/Single Key Valve
No. of Ports2 Ports
Stator MaterialPTFE/CTFE
Unit SizeEach