Designed for accurate pouring with minimal sample loss. Fisherbrand™ Polystyrene Diamond-Shaped Standard Weighing Boat features a smooth surface that flexes easily. Disposable boats have a stable, flat bottom and can be used in a variety of lab environments and applications.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Color
FKS12972850
100 mL
Black
FKS12962850
30 mL
Black
FKS12387552
100 mL
White
FKS11573422
30 mL
White
4 Options
Catalog number FKS12972850
Price (SGD)
176.00
PK
Add to cart
Capacity (Metric):
100 mL
Color:
Black
Price (SGD)
176.00
PK
Add to cart
Features a diamond shape that easily flexes for accurate pouring
Wide, flat bottom helps resist tipping
Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases
Temperature range -10 to 70°C
Can be used with microwave moisture analyzers
500 FDA/food-grade compliant boats per pack
Specifications
Bottom ShapeFlat
Capacity (Metric)100 mL
ColorBlack
DisposableSingle-use
MaterialPolystyrene
ShapeDiamond
TypeWeighing Boat
For Use With (Equipment)Microwave Moisture Analyzer