Hydrogen Generators
Hydrogen Generators
Actual product may vary
Fisherbrand
Hydrogen Generators
Research-grade hydrogen gas on demand
Have Questions?
Catalog NumberFlow RateModelWattage
FKS12965981250 mL/min.40H185w
Catalog number FKS12965981
Price (SGD)
48,559.00
EA
Add to cart
Flow Rate:
250 mL/min.
Model:
40H
Wattage:
185w
Price (SGD)
48,559.00
EA
Add to cart
Provide a safe and economical alternative to high-pressure gas cylinders and can be easily installed on the lab bench. Ideal for carrier and fuel gas applications and suitable for use with any laboratory instrument requiring a supply of ultra-high-purity hydrogen. Eliminate interruptions in analyses to change cylinders. Three models available with 160, 250 and 500cc/min. outlet flowrates.
  • LCD display shows system status and diagnostic capabilities
  • Automatic leak detection
  • Integrated microprocessor control
  • Low water and poor quality water detection and alarms
  • Built-in switch-mode power supply unit accepts electricity supplied between 100 and 230V and 50 to 60Hz
Products in this group have a universal power supply and can be used with any voltage 100-230V 50/60Hz
  • Produce a continuous stream of research-grade hydrogen gas from deionized water using a proprietary ion exchange membrane and drying cartridge
  • Using hydrogen as a GC carrier gas increases speed and sensitivity compared to helium, decreasing analysis time without discernible loss of resolution
Specifications
Certifications/ComplianceCE, CSA, UL
Depth (English)17.2 in.
Depth (Metric)34.2 cm
Flow Rate250 mL/min.
Height (English)17.9 in.
Height (Metric)45.6 cm
Max. Ambient Operating Temperature5°C to 40°C
Outlet Port0.125 in. compression fitting
Voltage90 to 264 V
Weight (Metric)23 kg
Width (English)13.5 in.
Electrical Requirements90 to 264V 50/60Hz
Model40H
Nitrogen Purity99.9995%
Pressure0 to 100 psi
Wattage185w
Warranty2 Years
Frequency50/60 Hz
TypeHydrogen Fuel Gas Generator
Unit SizeEA