Polypropylene Non-medicinal Sharps Container
Polypropylene Non-medicinal Sharps Container
Actual product may vary
Fisher Catalogue
Polypropylene Non-medicinal Sharps Container
Designed for use in public access areas such as A&E and out-patients. Sharpsafe™ Polypropylene Non-medicinal Sharps Container conforms to the UN standard covering the transport of dangerous goods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS129535251 L
FKS129135357 L
Catalog number FKS12953525
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1 L
  • UN approved
  • Sharpsguard
  • Rectangular shape
  • Simple to assemble
  • Yellow with orange lids
  • Conforms to BS 7320:1990, UN 3291,
  • Polypropylene for incineration or alternative treatment
Specifications
Capacity (Metric)1 L
Certifications/ComplianceBS 7320, AFNOR NX30-500, UN 3291
Closure TypeLid
ColorYellow, Orange
Model4160
Quantity1 Ea.
IncludesClosure
For Use With (Application)Designed for use in public access areas such as A&E and out-patients