Reduces the risk of damage to pipets during transportation and sterilisation. Nickel Electro™ Metalware Range™ Stainless Steel Square Sterilization Cans withstand dry heat temperatures of up to 180°C and can be steam sterilized at 120°C.
Have Questions?
Catalog Number
Dimensions (L x W x H)
Height (Metric)
FKS12946986
70 x 70 x 210 mm
210 mm
Catalog number FKS12946986
Price (SGD)
195.00
EA
Add to cart
Dimensions (L x W x H):
70 x 70 x 210 mm
Height (Metric):
210 mm
Price (SGD)
195.00
EA
Add to cart
Popular square pipet cans with permanent cast silicone rubber pads in lids and bases reduce the risk of damage to pipets during transportation and sterilisation.
Manufactured from stainless steel.
Can be steam sterilized at 120°C.
Withstands dry heat temperatures of up to 180°C.
Square section design allows easy stacking for efficient use of space and prevents cans from rolling off benchtop.
Specifications
AutoclavableAutoclavable
MaterialStainless Steel
TypeSquare Pipet Can
IncludesPermanent cast silicone rubber pads inlids and bottoms