Eutech™ pH Buffer Tablets
Eutech™ pH Buffer Tablets
Actual product may vary
Eutech™ pH Buffer Tablets
Eutech™ pH Buffer Tablets
Fisherbrand
Eutech™ pH Buffer Tablets
Eutech™ offers pH buffer tablets for use with pH meters and electrodes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberpH
FKS1292343710
FKS126311254
FKS126411257
Catalog number FKS12923437
Price (SGD)
209.00
PK
Add to cart
pH:
10
Price (SGD)
209.00
PK
Add to cart
Eutech™ pH buffer tablets provide a quick, convenient way to prepare pH buffers from tablets for pH calibrations. Each box contains 100 pH buffer tablets.
Eutech pH buffer tablets are designed to provide a quick and simple way to prepare pH buffers from tablets. Mix 30 ml of deionized or distilled water with each tablet. When prepared correctly, the resulting pH buffer solution will have a tolerance of +/- 0.05 pH.
  • Tablets are individually packaged for long shelf life storage
  • Tablets are easy to prepare
  • Tablets are lightweight and compact, ideal for field use
Specifications
ColorGreen
For Use With (Application)pH calibration
For Use With (Equipment)pH Meter and Electrode
pH10
Physical FormTablet
Quantity100 Tablets
Solution TypeBuffer
Unit SizePK