Stuart™ HM250C Model Heating Mantle
Stuart™ HM250C Model Heating Mantle
Actual product may vary
Fisherbrand
Stuart™ HM250C Model Heating Mantle
Have Questions?
Catalog number FKS12920161
Price (SGD)
4,527.00
EA
Add to cart
Price (SGD)
4,527.00
EA
Add to cart
Specifications
Capacity (Metric)250 mL
Depth (Metric)175 mm
Dimensions (D x W x H) Interior175 x 175 x 150mm
Height (Metric)150 mm
Voltage230 V
Electrical Requirements230V, 50/60Hz, 200W
ModelHM250C
MaterialAluminum
ShapeRound
Wattage200w
Width (Metric)175mm
Product TypeHeating Mantle
Temperature (Metric)450°C
For Use With (Application)Emergency cut out which will automatically trigger in the event of a spillage