Soda Lime Glass Petri Dishes
Actual product may vary
Fisherbrand
Soda Lime Glass Petri Dishes
High-quality petri dish, with lid.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Height (Metric)
FKS1291140880 mm15 mm
FKS12921408100 mm15 mm
Catalog number FKS12911408
Price (SGD)
207.00
Pack
Diameter (Metric):
80 mm
Height (Metric):
15 mm
Price (SGD)
207.00
Pack
  • Constructed of soda lime glass.
  • Includes lid
Specifications
Closure IncludedClosure Included
MaterialSoda Lime Glass
ShapeRound
TypePetri Dish
Height (Metric)15 mm
Diameter (Metric)80 mm
Unit SizePack