Centrifuge 5430/5430 R
Centrifuge 5430/5430 R
Actual product may vary
Fisherbrand
Centrifuge 5430/5430 R
The unique, cross-over Eppendorf Centrifuge 5430 and 5430 R combine the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, they can accommodate rotors for microplates and 15/50 mL conical tubes as well as Eppendorf tubes and PCR strips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberElectrical RequirementsIncludesController TypeModel
FKS12893222230 V, 50/60 Hz (GB)Without rotorKeypadCentrifuge 5430 (EU-IVD)
FKS12853232230 V, 50/60 Hz (EU)With Rotor FA-45-30-11 incl. rotor lidRotary KnobCentrifuge 5430 (EU-IVD)
Catalog number FKS12893222
Price (SGD)
22,622.00
EA
Add to cart
Electrical Requirements:
230 V, 50/60 Hz (GB)
Includes:
Without rotor
Controller Type:
Keypad
Model:
Centrifuge 5430 (EU-IVD)
Price (SGD)
22,622.00
EA
Add to cart
  • Max. rotor capacity: 48 × 1.5/2.0 mL, 6 × 50 mL, 2 × MTP
  • Max. speed: 30,130 × g (17,500 rpm)
  • Remarkable versatility with 12 different rotors
  • Soft-touch one-finger lid closure for ergonomic operation
  • Menu-driven, multi lingual operation menu (English, German, French, Spanish) with large backlit display
  • 5 program keys for easy access to routine programs
  • Saves up to 50 user-defined programs
  • Automatic rotor recognition and imbalance detection for maximized operational safety
  • Eppendorf QuickLock™ – System for quickly opening and closing the rotor lid
  • Specifications
    Capacity30 x 1.5/2 mL
    Max. RCF30,130 x g
    RefrigeratedNo
    DisplayLarge, brightly lit LCD
    Height (English) Exterior9.8 in.
    Height (Metric) Exterior25 cm
    Noise Level<58 dB(A) with Rotor F-45-30-11
    Safety FeaturesLid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition
    Timer30 sec. to 99:59 hrs., with continuous run function
    Width (English) Exterior11.2 in.
    Width (Metric) Exterior34 cm
    Wattage475 W
    Certifications/ComplianceUL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
    TypeCentrifuge
    IncludesWithout rotor
    Electrical Requirements230 V, 50/60 Hz (GB)
    Controller TypeKeypad
    Depth (Metric)420 mm
    VentilatedYes
    Maximum Speed Range100 to 17,500 rpm
    Net Weight29 kg
    Run Time30 sec. to 9 hr. 59 min.
    StandardsEN ISO 14971, EN 61010-2-101, EN 61326-2-6, EN 62366, EN ISO 18113-1, EN ISO 18113-3, EN ISO 15223-1, EN 61010-1, EN 61010-2-020, IEC 61010-1, IEC 61010-2-020, EN 55011, EN 61326-1, EN 50581 (refer to manual for more details)
    Temperature Range2°C to 40°C
    Frequency50/60 Hz
    Voltage230 V
    ModelCentrifuge 5430 (EU-IVD)
    Unit SizeEA