Wooden Beaker Brushes
Wooden Beaker Brushes
Actual product may vary
Fisherbrand
Wooden Beaker Brushes
Features center set bristles. Pinnacle Brush™ Tube Wooden Beaker Brushes offer gentle cleaning for beakers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS12891924Brush Circumference: 18 cm
FKS1280193480 x 50 x 150 mm
Catalog number FKS12891924
Price (SGD)
89.00
PK
Add to cart
Size:
Brush Circumference: 18 cm
Price (SGD)
89.00
PK
Add to cart
  • Rounded shape
  • Wood handle
Specifications
Product TypeBeaker Brush
Bristle MaterialFiber
Handle MaterialWood
Diameter (Metric)5.7 cm
For Use With (Equipment)Beaker
Length (Metric)41 cm
Length (Metric) Brush15.5 cm
Length (Metric) Handle25.5 cm
Quantity6 Pk.
SizeBrush Circumference: 18 cm
Unit SizePK