Clear Soda Lime Glass Bottle with Fitted Closure
Clear Soda Lime Glass Bottle with Fitted Closure
Actual product may vary
Fisherbrand
Clear Soda Lime Glass Bottle with Fitted Closure
Use the Fisherbrand™ Soda Lime Glass Bottle with 28mm Fitted Closure, Clear in the laboratory to store and manipulate laboratory media for experimentation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1289088460 mL
FKS12800894100 mL
FKS12830894250 mL
FKS12840894300 mL
FKS12850894500 mL
FKS128608941000 mL
Catalog number FKS12890884
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
60 mL
  • Tamper evident vistop closure with polycone liner
  • ISO certified
  • Clear soda lime glass
Specifications
Capacity (Metric)60 mL
ColorClear
Diameter (Metric) Inner38.6 mm
MaterialGlass (Type 3)
Neck ID (Metric)28mm
No. per Pack40Pack
Quantity40Pack
Closure ColorWhite
Liner MaterialLDPE
Closure IncludedYes
ShapeRound
Height (Metric)94 mm
Product TypeLaboratory Bottle
Closure MaterialHDPE
Diameter (Metric)38.6 mm
Closure TypeTamper-evident Vistop
Unit SizePK