Disposable Inoculation Loop, 1μL
Product Image
Fisherbrand
Disposable Inoculation Loop, 1μL
Manufactured from polystyrene for strength and flexibility. Microspec™ Disposable Inoculation Loops are ideal for the cultivation of microbes on plates by transferring inoculum for streaking and to transfer microscopic organisms.
Have Questions?
Catalog NumberVolume
FKS128901551 μL
Catalog number FKS12890155
Price (SGD)
150.00
PK
Add to cart
Volume:
1 μL
Price (SGD)
150.00
PK
Add to cart
  • High impact, polystyrene, disposable inoculating loops
  • 200mm long to reduce the risk of contamination
  • Includes 50 grip seal bags of 20 ea.
Specifications
DisposableSingle-use
Length (Metric)200 mm
MaterialPlastic
SterilitySterile
Volume1 μL
Product TypeInoculating Loop
For Use With (Equipment)Laboratory and Research use
Quantity1000 Plastic Culture Loops (50 bags of 20)
For Use With (Application)Laboratory and Research use
Unit SizePK