Sterilin™ Disposable Glass Containers
Sterilin™ Disposable Glass Containers
Actual product may vary
Fisherbrand
Sterilin™ Disposable Glass Containers
Ideal for small volume samples, Thermo Scientific™ Sterilin™ Disposable Glass Containers are made from soda glass with a leakfree polypropylene cap.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1288279330 mL
Catalog number FKS12882793
Price (SGD)
-
Capacity (Metric):
30 mL
Ideal for small volume samples, Thermo Scientific™ Sterilin™ Disposable Glass Containers are made from soda glass with a leakfree polypropylene cap.
NOT AVAILABLE IN AMERICA
  • Soda glass offers superior chemical resistance to plastic but is not recommended for autoclaving
  • Tray packed for safety and convenience
  • Supplied with cap—replacement caps available for the 30mL universal
  • Supplied unlabeled and nonsterile
  • In vitro use only
Ordering Information:
  • Not available in all geographic regions
Note: This glassware is manufactured from Soda glass and therefore it is recommended that the product is not autoclaved.
Specifications
Closure IncludedYes
MaterialSoda Lime Glass
SterilityNon-sterile
Capacity (Metric)30 mL
Closure MaterialPolypropylene
DescriptionDisposable Glass Container
No. per Case375/Cs.
Unit SizePK