Replacement Clip for Cole-Parmer™ TR-200 Series Tube Rotators
Replacement Clip for Cole-Parmer™ TR-200 Series Tube Rotators
Actual product may vary
Fisherbrand
Replacement Clip for Cole-Parmer™ TR-200 Series Tube Rotators
Replacement Clip for Cole-Parmer™ TR-200 Series Tube Rotators replaces broken or weakened micro tube clips for the Stuart SB3/1 Tube Holder. Holds 10 to 11.5 mL micro tubes in place.
Have Questions?
Catalog number FKS12869435
Price (SGD)
259.00
PK
Add to cart
Price (SGD)
259.00
PK
Add to cart
  • Choose the tube holder(s) that accommodates your tube size(s).
  • Spare clips for microcentrifuge tube holder disc 00077-11.
  • 1 year warranty.
Specifications
Product TypeSpare Clip
For Use With (Equipment)SB3/1 Tube Holder
Unit SizePK