1x1 image pixel for data collection
High-Performance Uv Variable Intensity Transilluminator
High-Performance Uv Variable Intensity Transilluminator
Actual product may vary
Fisherbrand
High-Performance Uv Variable Intensity Transilluminator
Have Questions?
Catalog NumberWavelength
FKS12863928302 nm
Catalog number FKS12863928
Price (SGD)
11,930.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Wavelength:
302 nm
Price (SGD)
11,930.00
EA
Add to cart
UVP's high performance transilluminators feature 25W tubes and a specially constructed filter that provides the most uniform light emission possible from a tube based unit.
The units have a stainless steel top that is easy to clean and a hinged UV blocking lid to protect the user. High, medium and low intensity settings make the unit suitable for both visualisation and band cutting. An internal fan improves the tube lifetime.Dimensions: 337mm x 486mm x 143mm
Specifications
Wavelength302 nm
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link