Polypropylene Cryoboxes with Slip Lid
Polypropylene Cryoboxes with Slip Lid
Actual product may vary
Fisher Catalogue
Polypropylene Cryoboxes with Slip Lid
Use these polypropylene boxes with grid inserts for storage of up to 100 tubes. Fisherbrand™ Polypropylene Cryoboxes are ideal for sample storage and transportation.
Have Questions?
Catalog NumberColorTypeTube Array
FKS12861593NaturalCryo Box10 x 10
Catalog number FKS12861593
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Natural
Type:
Cryo Box
Tube Array:
10 x 10
  • Cryoboxes with Slip Lid, PP, Format 133 x 133mm
  • High quality polypropylene (PP)
  • Available in natural and other colors
  • Boxes are adjustable in height
  • The boxes are available in 6 modern colors, with an numerical coding and corresponding vents for draining condensed water
  • The 6 x 6 until 10 x 10 grid accommodates space from 36 to 100 tubes
  • autoclavable, temperature-resistant from -90°C to +121°C
  • Available grids: 6 x 6, 7 x 7, 8 x 8, 9 x 9, 10 x 10
  • Numerically coded
Specifications
ColorNatural
MaterialPolypropylene
TypeCryo Box
IncludesSlip Lid
Tube Array10 x 10
Length (Metric)133 mm
Width (Metric)133 mm
Height (Metric)50 to 75 mm
Unit SizePK