An excellent solution for heating, sterilization and combustion in the laboratory. R&L Enterprises™ Universal Bunsen Burners are designed to use a wide range of gases.
Have Questions?
Catalog Number
Height (Metric)
FKS12852144
160 mm
Catalog number FKS12852144
Price (SGD)
141.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Height (Metric):
160 mm
Price (SGD)
141.00
EA
Add to cart
For use with coal gas, natural (methane) gas, butane, propane or mixtures of these gases
With adjustable jet, screwed air regulator, gas control needle valve and adjustable baffle
Manufactured in brass with nickel plating
Gas sealant grease is applied to the thread of the tube and jet
Constructed using a heavy pressure diecast base painted in chemically resistant blue acrylic finish
Tested for flame quality and leakage prior to packing