Check Order Status
Nitrate Standard Solution Concentrate, 2ppm, Reagecon™
Element Search
Structure Search
Nitrate Standard Solution Concentrate, 2ppm, Reagecon™
Have Questions?
Catalog Number
Quantity
FKS12832503
100 mL
Catalog number
FKS12832503
Price (SGD)
375.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (SGD)
375.00
EA
Add to cart
Specifications
Packaging
100mL
Concentration
2 ppm
Quantity
100 mL