DURAN™ GLS 80™ O-Ring Sealing Gasket, Black EPDM rubber, GLS 80™ Wide Mouth
Product Image
Fisherbrand
DURAN™ GLS 80™ O-Ring Sealing Gasket, Black EPDM rubber, GLS 80™ Wide Mouth
Optional sealing gasket for GLS 80™ bottles.
Have Questions?
Catalog number FKS12826573
Price (SGD)
47.00
PK
Add to cart
Price (SGD)
47.00
PK
Add to cart
EPDM O-Ring gasket improves sealing between GLS 80™ stirred reactor and the glass bottles.
  • Improved sealing with GLS 80™ bottles
Specifications
For Use With (Application)Liquid tight seal
Quantity5/Pk.
Diameter (Metric) Outside78 mm
MaterialEPDM
Unit SizePK