Sterilin™ Silicone Stoppers
Sterilin™ Silicone Stoppers
Actual product may vary
Fisherbrand
Sterilin™ Silicone Stoppers
Guarantee a tight, reliable seal for your glass and plastic flasks. Suitable for use in various applications.
Have Questions?
Catalog NumberClosure SizeDiameter (Metric) BaseDiameter (Metric)TopNo. per Pack
FKS1282291310 mm10 mm12.5 mm10
Catalog number FKS12822913
Price (SGD)
187.00
PK
Add to cart
Closure Size:
10 mm
Diameter (Metric) Base:
10 mm
Diameter (Metric)Top:
12.5 mm
No. per Pack:
10
Price (SGD)
187.00
PK
Add to cart
Guarantee a tight, reliable seal for your glass and plastic flasks. Suitable for use in various applications. Thermo Scientific™ Sterilin™ Silicone Stoppers are suitable for use over a wide temperature range: -50° to +200°C.
  • Suitable for sealing glass and plastic flasks
  • Suitable for use over a wide temperature range: -50° to +200°C
  • Autoclavable
Compliance
  • Material from which the product is made is compliant with FDA-CFR 21 Part 177.2600 Food Contact
Ordering Information
  • Additional sizes available. Please contact Customer Service for more details. Not available in all geographic regions.
Specifications
Diameter (Metric) Base10 mm
Diameter (Metric)Top12.5 mm
MaterialSilicone
Length (Metric)20 mm
No. per Pack10
Temperature Range (Metric)-50°C to +200°C
AutoclavableAutoclavable
SterilityNon-sterile
Closure Size10 mm
Unit SizePK