Stainless Steel Chemical Spoon
Stainless Steel Chemical Spoon
Actual product may vary
Fisher Catalogue
Stainless Steel Chemical Spoon
Stainless Steel Chemical Spoon with double end is 150 mm long.
Have Questions?
Catalog NumberLength (Metric)
FKS12811431150 mm
Catalog number FKS12811431
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
150 mm
  • Material: Stainless Steel
  • Double Ended
  • Length: 150 mm
Specifications
Quantity1 Ea.
SterilityNon-sterile
AutoclavableAutoclavable
ShapeDouble Spoon
Product TypeChemical Spoon Double End Spatula
For Use With (Application)For Wide Laboratory Use
Length (English)5.9 in.
MaterialStainless Steel (304)
Length (Metric)150 mm
Unit SizeEA