Soda Lime Glass Bottle with 28mm Fitted Closure, Amber
Soda Lime Glass Bottle with 28mm Fitted Closure, Amber
Actual product may vary
Fisher Catalogue
Soda Lime Glass Bottle with 28mm Fitted Closure, Amber
Use the Fisherbrand™ Soda Lime Glass Bottle with 28mm Fitted Closure, Amber in the laboratory to store and manipulate laboratory media for experimentation.
Have Questions?
Catalog NumberCapacity (Metric)
FKS12810954500 mL
Catalog number FKS12810954
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
  • Tamper evident vistop closure with polycone liner
  • ISO certified
  • Amber soda lime glass
Specifications
Capacity (Metric)500 mL
ColorAmber
Diameter (Metric) Inner97.5 mm
MaterialGlass (Type 3)
Neck ID (Metric)28mm
No. per Pack10 Pack
Quantity10 Pack
Closure ColorWhite
Liner MaterialLDPE
Certifications/ComplianceISO
Closure IncludedYes
ShapeRound
Height (Metric)215.6 mm
Product TypeLaboratory Bottle
Closure MaterialHDPE
Diameter (Metric)97.5 mm
Unit SizePK