Soda Glass Capillary Tube
Soda Glass Capillary Tube
Actual product may vary
Fisherbrand
Soda Glass Capillary Tube
Use these tubes for melting point determinations. Soda Glass Capillary Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.
Have Questions?
Catalog NumberLength (Metric) TubeProduct Type
FKS12807685100 mmCapillary Tube
Catalog number FKS12807685
Price (SGD)
23.00
PK
Add to cart
Length (Metric) Tube:
100 mm
Product Type:
Capillary Tube
Price (SGD)
23.00
PK
Add to cart
Use these tubes for melting point determinations. Soda Glass Capillary Tubes provide excellent sample retention during centrifugation, and fill rapidly without affecting specimen integrity.
  • Soda glass capillary tubes
  • Outside diameter 1.8/2.0mm
  • Wall thickness 0.28/0.32mm
  • Packed in tubular cartons of 100
Specifications
Product TypeCapillary Tube
Length (Metric) Tube100 mm
Unit SizePK