Clinitrol™ 290 Reference Solution; Advanced Instruments
Clinitrol™ 290 Reference Solution; Advanced Instruments
Actual product may vary
Clinitrol™ 290 Reference Solution; Advanced Instruments
Critical normal human serum midpoint of 290mOsm
Have Questions?
Catalog NumberFor Use With (Equipment)Quantity
FKS128022012020, 3300 and 3320 Osmometers10 x 2 mL
Catalog number FKS12802201
Price (SGD)
-
For Use With (Equipment):
2020, 3300 and 3320 Osmometers
Quantity:
10 x 2 mL
Critical normal human serum midpoint of 290mOsm
  • Checks performance of freezing-point osmometers in the critical area of normal serum values
  • Contains known and controlled quantities of some of the same solutes which comprise osmotically active particles of serum
  • Pipetted into a tube and run just like normal serum
  • Runs as the first and last samples in a series of tests to monitor operator technique and instrument performance
  • Does not require mixing, reconstituting or refrigerating; one-year shelf life
Specifications
Certifications/ComplianceCE
ColorClear
Concentration290 mOsm/kg H2O
For Use With (Equipment)2020, 3300 and 3320 Osmometers
Sample TypeSerum
TypeReference Solution
IncludesSodium Chloride (NaCl) and 2-Hydroxybiphenyl Sodium Salt Tetrahydrate
Intended UseOsmometer instrument calibration and quality control
Quantity10 x 2 mL
Shelf Life3 Years
FormLiquid
Unit SizeEA
Contents & Storage
2°C to 30°C