Dissecting Board
Dissecting Board
Actual product may vary
Dissecting Board
Dissecting Board
Dissecting Board
Dissecting Board
Fisherbrand
Dissecting Board
No swelling, warping or cracking when you use Fisherbrand™ Richard-Allan Scientific™ Dissecting Boards.
Have Questions?
Catalog NumberWidth (English)
FKS1277086611 in.
Catalog number FKS12770866
Price (SGD)
-
Width (English):
11 in.
No swelling, warping or cracking when you use Thermo Scientific™ Richard-Allan Scientific™ Dissecting Boards, crafted of high-density polyethylene.
Features:
  • Dimensional scales permit quick sizing and serve as part of permanent record
  • Rubber feet stabilize working surface and help prevent board from slipping on workbench
  • White background contrasts with specimens for excellent photographic detail
  • Boards have a spill-containment border
  • Styrene-capped pins secure specimens and are available separately (09-002-18)
Dimensions:
  • Large board measures: 23 x 16 x 0.38 in. (58.4 x 40.6 x 0.97cm)
  • Small board measures: 13 x 11 x 0.63 in. (33 x 28 x 1.6cm)
Specifications
Product TypeDissecting Board
ColorWhite
Description1 Ea.
MaterialHDPE
Thickness (English)0.625 in.
Thickness (Metric)1.6 cm
Length (English)13 in.
Length (Metric)33 cm
Width (English)11 in.
Width (Metric)28 cm
Unit SizePK