Drying and Warming Cabinets
Drying and Warming Cabinets
Actual product may vary
Fisherbrand
Drying and Warming Cabinets
The Genlab™ Drying/Warming Cabinets are suitable for drying a range of items such as glassware and instruments and any general purpose warming applications.
Have Questions?
Catalog NumberCapacity (Metric)Safety FeaturesConvection TypeDimensions (L x W x H)
FKS12766095125 LSliding, toughened glass doorsNatural42 x 74 x 60 cm
Catalog number FKS12766095
Price (SGD)
5,669.00
EA
Add to cart
Capacity (Metric):
125 L
Safety Features:
Sliding, toughened glass doors
Convection Type:
Natural
Dimensions (L x W x H):
42 x 74 x 60 cm
Price (SGD)
5,669.00
EA
Add to cart
  • Non-insulated
  • Maximum temperature of 50°C above ambient
  • Thermostat control for set temperature
  • Fan circulation on 500 and 1000L models
  • Full two-year warranty
  • C.E. compliant
  • Exterior constructed from sheet steel finished in an easy clean powder coated paint
  • Fitted with adjustable shelf runners, removable plated wire grid shelves and ventilation outlets
  • The 125 and 250L models have sliding, toughened glass doors; the 500 and 1000L models have hinged, toughened glass doors
  • Control is via a direct reading thermostat with calibrated scale and tamper-proof lock
  • Mains switch with indicator and heat indicator
Specifications
Certifications/ComplianceC.E. compliant
Controller TypeDirect reading thermostat with calibrated scale and tamper-proof lock
Convection TypeNatural
Capacity (Metric)125 L
Temperature (Metric)Ambient to 50°C
TypeDrying Cabinet
For Use With (Application)Drying a range of items such as glassware and instruments and any general purpose warming applications
ModelDC125
No. of Shelves3
Safety FeaturesSliding, toughened glass doors
Warranty2 Years
Dimensions (D x W x H) Interior40 x 72 x 43 cm
Dimensions (L x W x H)42 x 74 x 60 cm
Electrical Requirements230 V 50/60 Hz