BioClean™ N-Plus BNPS 16 in. Sterile Nitrile Cleanroom Gloves
Actual product may vary
Fisherbrand
BioClean™ N-Plus BNPS 16 in. Sterile Nitrile Cleanroom Gloves
Elbow-length nitrile gloves offer resistance to a range of chemicals and are hand specific to enable prolonged use. Recommended for Class 10/ISO 4 critical environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS127658457.5
FKS127458456.5
FKS127558457
Catalog number FKS12765845
Price (SGD)
-
Size:
7.5
Processed in a cleanroom environment to ensure compatibility with ISO Class 4 operating conditions
  • Extra-length nitrile cleanroom gloves
  • Sterilzed by gamma irradiation
  • Textured surface, powder free, beaded cuff
  • Hand-specific and anatomically shaped
  • Extended leaching in pure deionized water ensures low levels of chlorides and extractable metals
  • AQL: 0.65 for holes (1000mL water test)
  • Compatible for use in ISO Class 4
  • USP 797 Compliant
  • USP 800 Compliant
  • ASTM D6978 05 Tested
  • Double Donnable
Specifications
Certifications/ComplianceASTM D6978-05
Certifications/Compliance (Europe)CE 0598, EN 420:2003 + A1:2009, EN 421:2010, EN 455 Part 2, EN ISO 374-1:2016, EN ISO 374-5:2016, Food Contact, Category III
Cleanroom ClassClass 10 (ISO 4)
AQL0.65
ColorWhite
DisposableSingle-use
Exterior FinishFully Textured
Finger Thickness (English)7.8 mil
Finger Thickness (Metric)0.20 mm
HandSpecific
Latex FreeYes
MaterialNitrile
Palm Thickness (English)6.6 mil
Palm Thickness (Metric)0.17 mm
PowderedPowder Free
Product TypeCleanroom Gloves
Size7.5
SterilitySterile
Thickness (English)7.8 mil
Thickness (Metric)0.20 mm
Manufacturing/QMS Audit StandardsISO 9001, PPE Regulation 2016 425 Module D
ModelBNPS75
SeriesBioClean™ N-Plus BNPS
Length (English)16 in.
Length (Metric)40 cm
Unit SizePack