Use this dissecting kit for a variety of applications. Dissecting Sets in Plastic Wallets are suitable for theatre, ward, veterinary, dental and chiropody use.
Have Questions?
Catalog Number
Material
FKS12764416
Stainless Steel
Catalog number FKS12764416
Price (SGD)
105.00
EA
Add to cart
Material:
Stainless Steel
Price (SGD)
105.00
EA
Add to cart
Basic, student quality stainless steel instruments in wallet
Set includes:
1 forceps (125mm, straight, fine point)
1 scalpel (blade length 38mm)
1 forceps (125mm, straight, blunt tip)
1 scissors (125mm straight, fine point)
2 needles in metal handle
Specifications
Case MaterialPlastic
Includes1 forceps (125mm, straight, fine point), 1 scalpel (blade length 38mm), 1 forceps (125mm, straight, blunt tip), 1 scissors (125mm straight, fine point), 2 needles in metal handle