WypAll™ L10 General Clean™ Light Duty Wipes, Centrefeed Roll
Actual product may vary
WypAll™ L10 General Clean™ Light Duty Wipes, Centrefeed Roll
WypAll™ L10 General Clean™ Light Duty Wipes, Centrefeed Roll
WypAll™ L10 General Clean™ Light Duty Wipes, Centrefeed Roll
WypAll™ L10 General Clean™ Light Duty Wipes, Centrefeed Roll
WypAll™ L10 General Clean™ Light Duty Wipes, Centrefeed Roll
WypAll™ L10 General Clean™ Light Duty Wipes, Centrefeed Roll
WypAll™ L10 General Clean™ Light Duty Wipes, Centrefeed Roll
Fisherbrand
WypAll™ L10 General Clean™ Light Duty Wipes, Centrefeed Roll
Food and hygiene wiping paper delivers exceptional strength and absorbency for light-duty wiping. Centrefeed roll cleaning wipes are compatible with the WypAll™ Reach PLUS™, and Aquarius™ Roll Control™ wiper dispensers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging Quantity
FKS127643866 Rolls x 630 Sheets
FKS127543866 Rolls x 525 Sheets
Catalog number FKS12764386
Price (SGD)
-
Packaging Quantity:
6 Rolls x 630 Sheets
  • Delivered in a centrefeed roll format
  • Versatile mid-size wiper rolls can be used inside or outside a dispenser and offer more sheets per roll, resulting in less refills and fewer trucks on the road
  • Roll compatible with wiper dispensers and ReachPlus™ dispenser (6521), offering more reliable single sheet dispensing solutions
  • Unique Airflex™ technology deliver exceptional strength and absorbency
  • Perforated centrefeed rolls are suitable for food contact and ideal for light duty hygiene wiping such as cleaning food preparation areas
Specifications
Product TypeWiping Paper
Length (English)15.35 in.
Length (Metric)58.5 cm
Width (English)15.35 in
Width (Metric)39 cm
MaterialPaper
ColorBlue
Dimensions (L x W)38 x 18.3 cm (15 x 7.2 in.)
FormatCenterfeed Roll
For Use With (Application)For everyday cleaning, frequent light wiping and small spills
For Use With (Equipment)Aquarius wiper dispenser
Packaging Quantity6 Rolls x 630 Sheets
Ply1-ply