Check Order Status
Aluminum Foil Weighing Boats
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Fisherbrand
Aluminum Foil Weighing Boats
Have Questions?
Contact Us
Catalog Number
Height (Metric)
FKS12763355
6 mm
Catalog number
FKS12763355
Price (SGD)
-
Contact Us
Height (Metric):
6 mm
Specifications
Bottom Shape
Flat
Material
Aluminum
Shape
Square
Type
Weighing Boat
Size
2
Thickness
0.035 (Foil Gauge) mm
Length (Metric)
22 mm
Width (Metric)
6 mm
Height (Metric)
6 mm
Depth (Metric)
6 mm
Weight (Metric)
72 mg
Unit Size
PK
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
0 results displayed, search above for a specific certificate
Request a Certificate
Share catalog number, name or link