Labtek™ Liquid Detergent
Product Image
Fisherbrand
Labtek™ Liquid Detergent
Labtek liquid is a free-rinsing liquid alkaline detergent, for cleaning laboratory instruments, glassware, ceramics and plastics
Have Questions?
Catalog NumberVolume (Metric)
FKS127618755 L
Catalog number FKS12761875
Price (SGD)
-
Volume (Metric):
5 L
Labtek liquid is a free-rinsing liquid alkaline detergent, for cleaning laboratory instruments, glassware, ceramics and plastics. Labtek liquid is free from enzymes and sodium hypochlorite. Used in a 2% to 10% solution it rapidly removes oil, grease, tar, silicones, inorganic soils, polyacrylate and PVA resins, blood, proteins and a wide range of other organic soils from ferrous metals, glassware, plastics and ceramics. Can be used manually or in an ultrasonic bath where heating the solution improves cleaning efficiency. Not suitable for use on non-ferrous metals and polycarbonate.
  • Multi purpose
  • Powerful wetting agent
  • Ultrasonic compatible
  • Free rinsing The product is highly suitable for use in pharmaceutical, scientific and industrial laboratories where detergent residues are an issue.
Specifications
Product TypeLabtek Liquid Detergent
For Use With (Application)Suitable for use in pharmaceutical, scientific and industrial laboratories where detergent residues are an issue
Quantity1 Ea.
Volume (Metric)5 L
Unit SizeEA