Check Order Status
Borosilicate Glass Tall Form Beakers
Actual product may vary
Fisherbrand
Borosilicate Glass Tall Form Beakers
Mix and measure liquid ingredients to yield experimental results using Borosilicate Glass Tall Form Beakers. The matte finish beakers have no spout and have graduation markings.
Have Questions?
Catalog Number
Capacity (Metric)
FKS12757806
400 mL
Catalog number
FKS12757806
Price (SGD)
181.00
PK
Add to cart
Capacity (Metric):
400 mL
Price (SGD)
181.00
PK
Add to cart
Graduated
Without spout
Tall form
Translucent borosilicate glass
Matte finish
Specifications
Diameter (Metric) Outer
70mm
Capacity (Metric)
400 mL
Material
Borosilicate Glass
Height (Metric)
130mm
Quantity
10Pack
Type
Beaker
Graduated
Yes
Style
Tall Form