Classic Autoclave
Classic Autoclave
Actual product may vary
Fisherbrand
Classic Autoclave
Have Questions?
Catalog NumberCapacity (Metric)
FKS127553459 L
Catalog number FKS12755345
Price (SGD)
7,168.00
EA
Add to cart
Capacity (Metric):
9 L
Price (SGD)
7,168.00
EA
Add to cart
The availability of a wide range of model sizes, features and internal furniture options make the Prestige Medical Classic a popular choice where a cost effective, portable autoclave is needed. Fully automatic, simply press the button to start. Lightweight and portable, ideal for a variety of sterilisation solutions.
  • High quality construction
  • Fully automatic electronic time and temperature control
  • Steam release valve automatically opens to release excess pressure
  • Automatic fault warning light and cut-out if unit boils dry or sterilisation is interrupted
  • Pressure and temperature gauges
  • Complete with instrument basket
  • CE marked
  • Comprehensive one year warranty (UK)
  • Automatic electronic time and temperature control with end of cycle cut off and buzzer
  • Indicator lights for power on, heating, sterilisation cycle, fault condition and sterilisation complete
  • Incorrectly fitted lid does not allow pressure to build up
  • Steam release valve automatically releases excess pressure
  • Safety fuse operates if temperature is too high
  • If water level is too low or sterilisation cycle is interrupted a red fault lights operates and the autoclave switches off
  • DoH performance validated model with thermocouple entry port, pressure and temperature gauge For the sterilisation of unwrapped solid instruments and devices.
Specifications
Capacity (Metric)9 L
Unit SizeEA