B114 Disposable Deioniser
B114 Disposable Deioniser
Actual product may vary
Fisherbrand
B114 Disposable Deioniser
The B114 is the smallest unit in the Elga™ range of deionisers, designed to meet the needs of users who require small amounts of high purity water.
Have Questions?
Catalog number FKS12720916
Price (SGD)
6,653.00
EA
Add to cart
Price (SGD)
6,653.00
EA
Add to cart
  • Produces up to 30L/hr of deionised water
  • User is able to check water quality at the press of a button using the battery operated meter
  • Replacement cartridges can be fitted in seconds making the B114 quick and easy to maintain
Specifications
Capacity (Metric)Output of 20 L/cartridge to 250 L/cartridge (dependent on feed water)
DescriptionOutput, L/cartridge: 20 to 250 (dependant on feed
Flow Rate30 L/hr.
TypeWater Deionizer
Feedwater Pressure0.63 bar
For Use With (Application)Producing small amounts of high purity water
For Use With (Equipment)Cartridge number C114
Operating PressureInlet pressure (maximum), bar or PSIG or kg/cm2 : 0.63
Purity1MΩ/cm to 10MΩ/cm
Width (Metric)180 mm
Height (Metric)380 mm
Diameter (Metric)152 mm
Electrical RequirementsBattery operated
Unit SizeEA