Clifton™ aluminum Boiling Bath
Clifton™ aluminum Boiling Bath
Actual product may vary
Fisher Catalogue
Clifton™ aluminum Boiling Bath
Regulates from a gentle simmer to a vigorous boil, fitted with concentric ringed lid to vary the hole size. Nickel Electro™ Clifton™ Aluminum Boiling Bath produces steam that provides an effective source of heat for samples that may be damaged by direct flame.
Have Questions?
Catalog NumberModelController TypeTemperature Range
FKS127088781105 EREnergy regulator controls the amount of boiling from a gentle simmer to vigorous boilSimmer to boiling at 100°C
Catalog number FKS12708878
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Model:
1105 ER
Controller Type:
Energy regulator controls the amount of boiling from a gentle simmer to vigorous boil
Temperature Range:
Simmer to boiling at 100°C
Rectangular, aluminum electrically heated, multi-place baths regulates from a gentle simmer to a vigorous boil, fitted with concentric ringed lid to vary the hole size.
The steam produced provides an effective source of heat for samples:
  • that may be otherwise thermally damaged by a naked flame.
  • used for applications to distill/concentrate liquids, melt agar and other solids.
  • metallurgical analysis, protein analysis and bacteriological examinations
  • Features an integral constant level device to maintain constant water level during boiling and supplied with a set of rings to vary the diameter of aperture to accommodate different sizes of vessels of evaporating dishes and glassware, including round-bottom flasks.
  • Fitted with a sheathed immersion element and a one shot thermal cut out for safety.
Specifications
Controller TypeEnergy regulator controls the amount of boiling from a gentle simmer to vigorous boil
DescriptionBoiling Bath
Capacity (Metric)3.12 L
Temperature RangeSimmer to boiling at 100°C
For Use With (Application)Distilling/Concentrating Liquids, Melting Agar and Other Solids, Metallurgical Analysis, Protein Ana
MaterialAluminum
Model1105 ER
ShapeRectangular
Warranty2 Years
Dimensions (D x W x H)200 x 400 x 90 mm
Width (Metric)400 mm
Height (Metric)90 mm
Depth (Metric)200 mm
Weight (Metric)3.8 kg
Voltage230 V
Wattage1000 W
Unit SizeEA