Plastic Covers with Orifice for S 50 R Ultrasonic Cleaning Units
Plastic Covers with Orifice for S 50 R Ultrasonic Cleaning Units
Actual product may vary
Fisher Catalogue
Plastic Covers with Orifice for S 50 R Ultrasonic Cleaning Units
Allows glass beakers with a diameter between 90 and 97mm to be inserted into the bath for direct ultrasonic treatment. Fisherbrand™ Plastic Covers with Orifice for S 50 R Ultrasonic Cleaning Units can be combined with a rubber ring to securely and safely hold the beaker during the application.
Have Questions?
Catalog number FKS12699345
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Manufactured from durable blue plastic
Specifications
TypeLid
For Use With (Equipment)Ultrasonic Bath, S 50 R
MaterialPlastic
Unit SizeEA