Check Order Status
Polypropylene Pop-Up Tube Racks
Actual product may vary
Fisherbrand
Polypropylene Pop-Up Tube Racks
Opens and locks in place easily for quick use. Polypropylene Pop-Up Tube Racks provide space saving and compact storage for 15 and/or 50 mL tubes.
Have Questions?
Change view
Catalog Number
Color
Holds
FKS12693466
Purple
Tubes (15 mL), Tubes (17 mm)
FKS12603476
Green
Tubes (50 mL)
FKS11710634
Blue
Tubes (15 mL), Tubes (50 mL), Tubes (17 mm), Tubes (30 mm)
3 Options
Catalog number
FKS12693466
Price (SGD)
153.00
PK
Add to cart
Color:
Purple
Holds:
Tubes (15 mL), Tubes (17 mm)
Price (SGD)
153.00
PK
Add to cart
Made from polypropylene
Easy to open – easy to close
Designed to help organize 15 and 50 mL tubes
Imprinted grid aids quick retrieval
Stack racks in open or closed position
Saves space when racks are not in use
Not recommended for cleaning in autoclave
Specifications
Includes
Imprinted Grid
Product Type
Collapsible Tube Rack
Autoclavable
Not Autoclavable
Color
Purple
Holds
Tubes (15 mL), Tubes (17 mm)
Material
Polypropylene
Positions
45
Shape
Rectangular
Shelves
None
Stackable
Yes
Sterility
Non-sterile
For Use With (Equipment)
15 mL Test Tubes
Quantity
2 Pk.
Dimensions (L x W x H)
255 x 137 x 72 mm
Length (Metric)
255 mm
Length (English)
10 in.
Width (Metric)
137 mm
Height (Metric)
72 mm
Height (English)
2.8 in.
Diameter (Metric) Opening
17 mm
Weight (Metric)
1 kg