1x1 image pixel for data collection
Pyrex™ Borosilicate Glass Graduated Test Tubes with Stopper
Pyrex™ Borosilicate Glass Graduated Test Tubes with Stopper
Actual product may vary
Pyrex™ Borosilicate Glass Graduated Test Tubes with Stopper
Pyrex™ Borosilicate Glass Graduated Test Tubes with Stopper
Fisherbrand
Pyrex™ Borosilicate Glass Graduated Test Tubes with Stopper
Manufactured with borosilicate glass for excellent chemical and thermal shock resistance. Pyrex™ Borosilicate Glass Graduated Test Tubes are ideal for measuring and mixing liquids and solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)Capacity (Metric)
FKS1269304213 mm100 mm5 mL
FKS1136781426 mm200 mm50 mL
Catalog number FKS12693042
Price (SGD)
663.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Diameter (Metric):
13 mm
Length (Metric):
100 mm
Capacity (Metric):
5 mL
Price (SGD)
663.00
PK
Add to cart
  • White enamel graduations for visual volume assessment
  • Polyethylene stopper prevents spills and leakage
Specifications
Closure MaterialPolyethylene
Capacity (Metric)5 mL
Length (Metric)100 mm
MaterialBorosilicate Glass
IncludesInterchangeable stopper
Quantity10 Pack
GraduatedYes
Diameter (Metric)13 mm
Closure TypeStopper
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link