Universal Oven UN with Natural Convection & SingleDISPLAY
Universal Oven UN with Natural Convection & SingleDISPLAY
Actual product may vary
Universal Oven UN with Natural Convection & SingleDISPLAY
Universal Oven UN with Natural Convection & SingleDISPLAY
Fisherbrand
Universal Oven UN with Natural Convection & SingleDISPLAY
Ideal for drying, heating, ageing, burn-in and hardening in research, science, industry and quality assurance.
Have Questions?
Catalog NumberCapacity (Metric)No. of Shelves
FKS1268697753 L4
Catalog number FKS12686977
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
53 L
No. of Shelves:
4
Precise, easy to use, high quality - these three words describe the Memmert Universal Oven UN.
  • Model sizes: 30, 55, 75, 110, 160, 260, 450, 750
  • Temperature adjustment range: up to +300°C
  • Defined and programme-controlled fan speed
  • All-round heating system: absolutely uniform and precise temperature distribution
  • Pre-heated fresh air prevents temperature fluctuations
  • AtmoCONTROL software on a USB stick for programming, managing and transferring programmes via Ethernet interface or USB port
  • ControlCOCKPIT: TwinDISPLAY - Adaptive multifunctional digital PID-microprocessor controller with 2 high-definition TFT-colour displays.
  • adjustable parameters: temperature (Celsius or Fahrenheit), fan speed, air flap position, programme time, time zones, summertime/wintertime
  • Overtemperature protection
The technologically perfected masterpiece made of high-quality, hygienic, easy-to-clean stainless steel leaves nothing to be desired in terms of ventilation and control technology.
Specifications
Certifications/ComplianceCE, cUL, DVE, EAC, GS
Chamber MaterialStainless Steel (304/1.4301)
Controller TypeSingleDISPLAY. Adaptive multifunctional digital PID-microprocessor controller
Convection TypeNatural
Capacity (Metric)53 L
Temperature (Metric)20°C to 300°C
TypeHeating Oven
Display1 high-definition TFT-colour display (SingleDISPLAY)
For Use With (Application)Temperature Control in Science, Research and Material Tests in Industry
IncludesPC Software AtmoCONTROL
InsulationFully isolated Stainless Steel door with 2-point locking (compression door lock)
MaterialStainless Steel
Max. Shelf Load20 kg
ModelUN55
No. of Shelves4
Plug TypeMains cable with plug (German type)
Safety FeaturesTemperature control, autodiagnostic system
Shelf MaterialStainless Steel
Temperature ControlAdaptive multifunctional digital PID-microprocessor controller
Temperature StabilityUp to 99.9°C: 0.1 / from 100°C: 0.5
TimerDigital backwards counter with target time setting, adjustable from 1 minute to 99 days
Warranty3 years
Dimensions (D x W x H) Interior330 x 400 x 400 mm
Dimensions (L x W x H)585 x 784 x 514 mm
Length (Metric)514 mm
Width (Metric)585 mm
Width (Metric) Interior400 mm
Height (Metric)784 mm
Height (Metric) Interior400 mm
Depth (Metric)514 mm
Depth (Metric) Interior330 mm
Shipping Weight (Metric)67 kg
Weight (Metric)57 kg
Electrical Requirements115/230 V, 50/60 Hz
Voltage115/230 V
Wattage1700/2000 W