Lab Vision™ Ki-67, Rabbit Monoclonal Antibody
Lab Vision™ Ki-67, Rabbit Monoclonal Antibody
Actual product may vary
Lab Vision™ Ki-67, Rabbit Monoclonal Antibody
Lab Vision™ Ki-67, Rabbit Monoclonal Antibody
Fisherbrand
Lab Vision™ Ki-67, Rabbit Monoclonal Antibody
Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ Ki-67, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
Have Questions?
Catalog NumberFormQuantity
FKS12683697Concentrated100 μL
Catalog number FKS12683697
Price (SGD)
1,073.00
PK
Add to cart
Form:
Concentrated
Quantity:
100 μL
Price (SGD)
1,073.00
PK
Add to cart
Ki-67 is a nuclear protein, which is expressed in proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.
Host Species: Rabbit
Clone: SP6
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide derived from the human Ki-67 protein
Positive Control: Spleen, tonsil, or Breast CA
Cellular Localization: Nuclear
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
General References:
  1. Keszthelyi R et al. Immunomorphological Assessment of the New Ki-67 Specific Rabbit Monoclonal SP6 Antibody on Fixed-embedded Tissue Sections. Submitted for publication in Pathology Oncology Research.
Specifications
AntigenKi-67
ApplicationsImmunohistochemistry (Paraffin)
ClassificationMonoclonal
CloneSP6
ConjugateUnconjugated
Host SpeciesRabbit
ImmunogenA synthetic peptide derived from the human Ki-67 protein
Quantity100 μL
Regulatory StatusIVD
Research DisciplineCancer and Tumor Biology
Primary or SecondaryPrimary
Target SpeciesHuman
FormConcentrated
IsotypeIgG
Unit SizePK