PYREX™ Borosilicate Glass Graduated Cylinders
Actual product may vary
Fisherbrand
PYREX™ Borosilicate Glass Graduated Cylinders
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Tall Form provide superior chemical resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS126820421000 mL10 mL
FKS1264204250 mL1 mL
Catalog number FKS12682042
Price (SGD)
942.00
Pack
Capacity (Metric):
1000 mL
Graduations:
10 mL
Price (SGD)
942.00
Pack
Printed with blue enamel graduations for enhanced visibility. Pyrex™ Borosilicate Glass Cylinders in Tall Form provide superior chemical resistance.
  • Hexagonal base delivers stability
  • Heavy duty construction increases mechanical strength and reduces breakage (lowering cost and improving user safety)
  • Heavily beaded rim and spout allow clean and easy dispensing
  • Class B
  • Complies with ISO 4788
Specifications
Tolerance±10 mL
MaterialBorosilicate Glass
Certifications/ComplianceBS 604, DIN 12680, ISO 4788
ClassClass B
Capacity (Metric)1000 mL
Height (Metric)465 mm
ScaleBlue enamel
Graduations10 mL
Base TypeHexagonal
Scale ColorBlue
ShapeRound
Product TypeGraduated Cylinder
Unit SizePack