Search Thermo Fisher Scientific
Catalog Number | Length (Metric) | Volume (Metric) |
---|---|---|
FKS12679591 | 5.1 cm | 100 to 1000 μL |
FISF171103G | 3 cm | 0.1 to 10 μL |
FKS11921483 | 3 cm | 0.1 to 10 μL |
FKS12649591 | 5.1 cm | 10 to 100 μL |
FKS11912078 | 5.1 cm | 20 to 200 μL |