1000 μL Universal Pipetter Tips: Bevelled
1000 μL Universal Pipetter Tips: Bevelled
Actual product may vary
Fisher Catalogue
1000 μL Universal Pipetter Tips: Bevelled
Compatible with most popular brands of pipettes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityColorFormat
FKS12679535Non-sterileBlueRack
FIS14-222-690Non-sterileBlueBulk
Catalog number FKS12679535
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Sterility:
Non-sterile
Color:
Blue
Format:
Rack
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
Specifications
Volume (Metric)100 to 1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorBlue
AutoclavableAutoclavable
Clean ClaimsDNase, RNase, and Pyrogen free
FormatRack
Tip StyleBeveled, Universal
PackagingHinged rack
For Use With (Equipment)Universal Fit