MBL™ One Mark Class B Glass Pipets
MBL™ One Mark Class B Glass Pipets
Actual product may vary
Fisherbrand
MBL™ One Mark Class B Glass Pipets
Manufactured from soda lime glass with tooled jets to reduce breakage and complies with ISO 648 and DIN 12690. MBL™ One Mark Class B Glass Pipets incude markings in permanent amber stain which are diffused into the surface of the glass for increased chemical resistance. Calibrated for delivery.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ToleranceColor
FKS1267303220 mL±0.06 mLYellow
FKS12510026100 mL±0.15 mLYellow
FKS125600362 mL±0.02 mLOrange
Catalog number FKS12673032
Price (SGD)
127.00
PK
Add to cart
Volume (Metric):
20 mL
Tolerance:
±0.06 mL
Color:
Yellow
Price (SGD)
127.00
PK
Add to cart
  • Complies with ISO 648 and DIN 12690
  • Calibrated for delivery
  • Manufactured from soda lime glass with tooled jets to reduce breakage
  • Markings in permanent amber stain which are diffused into the surface of the glass for increased chemical resistance
Specifications
CalibrationTo Deliver
Certifications/ComplianceISO 648, DIN 12690
ClassB
MaterialSoda Lime Glass
GraduatedYes
Graduation ColorAmber
Quantity5 Pack
Tolerance±0.06 mL
ColorYellow
Volume (Metric)20 mL
TypeBulb Pipet
Unit SizePK