Scan 500 Automatic Colony Counter
Scan 500 Automatic Colony Counter
Actual product may vary
Fisherbrand
Scan 500 Automatic Colony Counter
Scan 500 is an automatic colony counter that ensures fast and efficient counting of colonies, including on chromogenic agar. It provides excellent reproducibility.
Have Questions?
Catalog NumberProduct Type
FKS12668685Automatic Colony Counter
Catalog number FKS12668685
Price (SGD)
60,443.00
EA
Add to cart
Product Type:
Automatic Colony Counter
Price (SGD)
60,443.00
EA
Add to cart
Automatic colony counter & inhibition zone reader, delivered with software.
  • Counting on Petri dishes (55 - 90 mm)
  • Spiral™ plating
  • Color detection: 7 colors on the same dish
  • Counting on chromogenic medium
  • Inhibition zone measurement
Specifications
Product TypeAutomatic Colony Counter
Connection TypeUSB
MagnificationCMOS color camera, M12 lens, Zoom x 28, Resolution: 1 megapixel
MaterialStainless Steel
Outlet StyleUSB
Total CountCounting time: up to 1000 colonies per second, Number of antibiotic paper disks: up to 7 antibiotics on a dia. 90 mm Petri dish, Reading time: 7 inhibition zone reading between 1 to 3 seconds
Dimensions (L x W x H)28.7 x 27.1 x 29 cm
Depth (English)10.67 in.
Height (English)11.42 in.
Width (English)11.30 in.
Weight (Metric)8.38 kg
Frequency50-60 Hz
Voltage100/240 V
WattageMax power: 20 W
Certifications/Compliance21 CFR Part 11, ISO 7218 and AOAC 977.27
For Use With (Application)The Scan 500 colony counter is used in the food, environmental, pharmaceutical, cosmetics, veterinary and public institutes research.
For Use With (Equipment)Counting on round Petri Dishes of dia. 55 - 90 mm, PetriFilmTM, MC-Media PadsTM, Compact DryTM, easyPlateTM, filtration membranes, Compatible with dataLinkTM or dataLinkTMpro,traceability system
Includes1 Scan™ software, 1 15 V power cord, 1 USB cable, 3 validation plates, 1 user manual
Unit SizeEA