Check Order Status
Plastic Slide Folders
Actual product may vary
Fisherbrand
Plastic Slide Folders
Easily color-code your slides with these plastic slide folders, available in a variety of colors.
Have Questions?
Change view
Catalog Number
Color
FKS12668326
Blue
FKS12608336
White
2 Options
Catalog number
FKS12668326
Price (SGD)
794.00
PK
Add to cart
Color:
Blue
Price (SGD)
794.00
PK
Add to cart
Hold up to 20 175 x 26mm ( in. x 1 in.) slides for convenient storage
Transparent top for easy identification of contents
Please Note: Availability may vary by country.
Specifications
Color
Blue
Device Holds
20 Slides
Length (English) Slide
3 in.
Length (Metric) Slide
75 mm
Material
Plastic
Stackable
Yes
Width (English) Slide
1 in.
Width (Metric) Slide
26 mm