PTFE Stir Bars with Pivot Ring
PTFE Stir Bars with Pivot Ring
Actual product may vary
Fisherbrand
PTFE Stir Bars with Pivot Ring
Provides stable stirring action. Fisherbrand™ PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
FKS1265115612 mm6 mm
FKS1001813160 mm10 mm
FKS1187886240 mm8 mm
FKS1150781225 mm6 mm
FKS1184886225 mm6 mm
FKS1183886220 mm6 mm
FKS1158780212 mm4.5 mm
FKS118088628 mm3 mm
FKS123495078 mm3 mm
Catalog number FKS12651156
Price (SGD)
36.00
PK
Add to cart
Length (Metric):
12 mm
Diameter (Metric):
6 mm
Price (SGD)
36.00
PK
Add to cart
Provides stable stirring action. FisherbrandTM PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.
  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action.
  • Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Diameter (Metric)6 mm
Length (Metric)12 mm
ShapeCylinder, Pivot ring
Unit SizePK